Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucoquinizarin 98.0+%, TCI America™
SDP

Catalog No. T0116500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0116500G 500 g
T011625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0116500G Supplier TCI America Supplier No. T0116500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 476-60-8
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00058948
InChI Key BKNBVEKCHVXGPH-UHFFFAOYSA-N
Synonym 2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone, 1,4-Dihydroxyanthrahydroquinone, leuco-Quinizarin, 1,4,9,10-Tetrahydroxyanthracene
PubChem CID 68063
IUPAC Name anthracene-1,4,9,10-tetrol
SMILES OC1=C2C(O)=C3C=CC=CC3=C(O)C2=C(O)C=C1

Specifications

Melting Point 151°C
Quantity 500 g
Formula Weight 242.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Leucoquinizarin
RTECSNumber OH4742000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.