accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4760-53-6

IUPAC Name: 2-(4-amino-2-chlorophenyl)-2-phenylacetonitrile
Synonyms: 2-(4-amino-2-chlorophenyl)-2-phenylacetonitrile
Molecular Weight (g/mol): 242.71
Molecular Formula: C14H11ClN2
InChi Key: ISOPQUKKXCXJIJ-UHFFFAOYNA-N
SMILES: NC1=CC=C(C(C#N)C2=CC=CC=C2)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results