Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chlorodiphenylacetonitrile, 95%, Thermo Scientific™

Catalog No. AC224370250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC224370250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224370250 Supplier Thermo Scientific Chemicals Supplier No. 224370250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4760-53-6
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C14H11ClN2
Quantity 25g
MDL Number MFCD00066975
Synonym 4-amino-2-chlorophenyl phenyl acetonitrile, 2-4-amino-2-chlorophenyl-2-phenylacetonitrile, 4-amino-2-chlorophenyl phenylacetonitrile, benzeneacetonitrile, 4-amino-2-chloro-alpha-phenyl, 2-4-amino-2-chlorophenyl-2-phenylethanenitrile, benzeneacetonitrile, 4-amino-2-chloro-.alpha.-phenyl, 4-amino-2-chloro-alpha-phenylbenzeneacetonitrile
InChI Key ISOPQUKKXCXJIJ-UHFFFAOYNA-N
SMILES NC1=CC=C(C(C#N)C2=CC=CC=C2)C(Cl)=C1
IUPAC Name 2-(4-amino-2-chlorophenyl)-2-phenylacetonitrile
Molecular Weight (g/mol) 242.71
PubChem CID 107335
Formula Weight 242.71
Percent Purity ≥94%
Chemical Name or Material 4-Amino-2-chlorodiphenylacetonitrile, 95%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-298-1
RTECSNumber CY1692900
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.