accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOHOHO

CAS RN 4767-03-7

IUPAC Name: 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Synonyms: 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: PTBDIHRZYDMNKB-UHFFFAOYSA-N
SMILES: CC(CO)(CO)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)