accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4769-97-5

IUPAC Name: 4-nitro-1H-indole
Synonyms: 4-nitro-1H-indole
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: LAVZKLJDKGRZJG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CNC2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)