accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 477-57-6

IUPAC Name: 9,20,21,25-tetramethoxy-15,30-dimethyl-7,23-dioxa-15,30-diazaheptacyclo[22.6.2.2³,⁶.1⁸,¹².1¹⁴,¹⁸.0²⁷,³¹.0²²,³³]hexatriaconta-3,5,8(34),9,11,18,20,22(33),24(32),25,27(31),35-dodecaene
Synonyms: tetrandrine
Molecular Weight (g/mol): 622.76
Molecular Formula: C38H42N2O6
InChi Key: WVTKBKWTSCPRNU-UHFFFAOYNA-N
SMILES: COC1=CC=C2CC3N(C)CCC4=CC(OC)=C(OC)C(OC5=CC6=C(CCN(C)C6CC6=CC=C(OC1=C2)C=C6)C=C5OC)=C34
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results