accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4783-86-2

ON

CAS RN 4783-86-2

IUPAC Name: 4-phenoxypyridine
Synonyms: 4-phenoxypyridine
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: OATKXQIGHQXTDO-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results