Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxypyridine 95.0+%, TCI America™
SDP

Catalog No. P14135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14135G Supplier TCI America Supplier No. P14135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4783-86-2
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.199
MDL Number MFCD00235157
InChI Key OATKXQIGHQXTDO-UHFFFAOYSA-N
PubChem CID 249651
IUPAC Name 4-phenoxypyridine
SMILES C1=CC=C(C=C1)OC2=CC=NC=C2

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenoxypyridine
EINECSNumber (5)-0723
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.