accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N(E)

CAS RN 4786-20-3

IUPAC Name: (2E)-but-2-enenitrile
Synonyms: crotononitrile
Molecular Weight (g/mol): 67.09
Molecular Formula: C4H5N
InChi Key: NKKMVIVFRUYPLQ-NSCUHMNNSA-N
SMILES: C\C=C\C#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)