Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotononitrile (pract) 98%, Thermo Scientific™

Catalog No. AC405782500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405782500 250 g
AC405780010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405782500 Supplier Thermo Scientific Chemicals Supplier No. 405782500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4786-20-3
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
InChI Key NKKMVIVFRUYPLQ-IHWYPQMZSA-N
Synonym z-2-butenenitrile, cis-crotononitrile, cis-cyanopropene, isocrotonic nitrile, 2-butenenitrile, z, z-crotononitrile, cis-2-butenenitrile, cis-1-propenyl cyanide, cis-beta-methylacrylonitrile, 2-butenenitrile
PubChem CID 643774
IUPAC Name (Z)-but-2-enenitrile
SMILES CC=CC#N

Specifications

Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.4180 to 1.4200 (20°C, 589nm)
Linear Formula CH3CHCHCN
Quantity 250 g
Formula Weight 67.09
Percent Purity ≥97.5% (mixture of cis and trans)
Physical Form Liquid
Chemical Name or Material Crotononitrile (pract), 98%
Hazard Category GHS Signal Word: Danger
EINECSNumber 225-335-1
RTECSNumber GQ6322000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.