CAS RN 478980-04-0
CAS RN 478980-04-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 1,3-Bis(2,4,6-Trimethylphenyl)Imidazol-2-Ylidene (1,4-Naphthoquinone)Palladium(0) Dimer
Sigma Aldrich 1,3-Bis(2,4,6-Trimethylphenyl)Imidazol-2-Ylidene (1,4-Naphthoquinone)Palladium(0) Dimer
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.