accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4792-83-0

IUPAC Name: 1,2-bis(4-ethoxyphenyl)-2-oxohydrazin-2-ium-1-ide
Synonyms: 1,2-bis(4-ethoxyphenyl)-2-oxohydrazin-2-ium-1-ide
Molecular Weight (g/mol): 286.33
Molecular Formula: C16H18N2O3
InChi Key: QUICZVHSJNKDBL-UHFFFAOYSA-N
SMILES: CCOC1=CC=C([N-][N+](=O)C2=CC=C(OCC)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results