accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONF

CAS RN 479253-00-4

IUPAC Name: tert-butyl 3-fluoropyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-fluoropyrrolidine-1-carboxylate
Molecular Weight (g/mol): 189.23
Molecular Formula: C9H16FNO2
InChi Key: SUECTKVSIDXQQE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(F)C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results