accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3Cl

CAS RN 479500-35-1

IUPAC Name: 1-butyl-1-methylpyrrolidin-1-ium chloride
Synonyms: 1-butyl-1-methylpyrrolidin-1-ium chloride
Molecular Weight (g/mol): 177.72
Molecular Formula: C9H20ClN
InChi Key: BOOXKGZZTBKJFE-UHFFFAOYSA-M
SMILES: [Cl-].CCCC[N+]1(C)CCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results