Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-1-methylpyrrolidinium Chloride 98.0+%, TCI America™
SDP

Catalog No. B33585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33585G 5 g
B335825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33585G Supplier TCI America Supplier No. B33585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 479500-35-1
Molecular Formula C9H20ClN
Molecular Weight (g/mol) 177.72
MDL Number MFCD08458922
InChI Key BOOXKGZZTBKJFE-UHFFFAOYSA-M
PubChem CID 11769095
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium chloride
SMILES [Cl-].CCCC[N+]1(C)CCCC1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 177.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.