CAS RN 480-40-0
Chrysin, 99.4%, For HPLC analysis, MP Biomedicals™
CAS: 480-40-0 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.24 MDL Number: MFCD00006593 InChI Key: RTIXKCRFFJGDFG-UHFFFAOYSA-N Synonym: 4-nitrophenyl-beta-d-glucopyranoside,pnpg,4-nitrophenyl beta-d-glucopyranoside,1-o-p-nitrophenyl-d-glucose,4-nitrophenyl beta-d-glucoside,p-nitrophenyl beta-d-glucopyranoside,p-nitrophenyl beta-d-glucoside,glucoside, p-nitrophenyl,p-nitrophenyl-beta-glucoside,p-nitrophenyl beta-glucoside PubChem CID: 92930 IUPAC Name: 5,7-dihydroxy-2-phenyl-4H-chromen-4-one SMILES: OC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
Chrysin, 99+%
CAS: 480-40-0 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.24 MDL Number: MFCD00006834 InChI Key: RTIXKCRFFJGDFG-UHFFFAOYSA-N Synonym: chrysin,5,7-dihydroxyflavone,chrysine,5,7-dihydroxy-2-phenyl-4h-chromen-4-one,crysin,4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl,unii-3cn01f5zj5,flavone, 5,7-dihydroxy,5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one,chembl117 PubChem CID: 5281607 ChEBI: CHEBI:75095 IUPAC Name: 5,7-dihydroxy-2-phenylchromen-4-one SMILES: C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O
5,7-Dihydroxyflavone, 98%
CAS: 480-40-0 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.241 MDL Number: MFCD00006834 InChI Key: RTIXKCRFFJGDFG-UHFFFAOYSA-N Synonym: chrysin,5,7-dihydroxyflavone,chrysine,5,7-dihydroxy-2-phenyl-4h-chromen-4-one,crysin,4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl,unii-3cn01f5zj5,flavone, 5,7-dihydroxy,5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one,chembl117 PubChem CID: 5281607 ChEBI: CHEBI:75095 IUPAC Name: 5,7-dihydroxy-2-phenylchromen-4-one SMILES: C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O
phyproof™ Chrysin, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Chrysin 98.0+%, TCI America™
CAS: 480-40-0 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.241 MDL Number: MFCD00006834 InChI Key: RTIXKCRFFJGDFG-UHFFFAOYSA-N Synonym: chrysin,5,7-dihydroxyflavone,chrysine,5,7-dihydroxy-2-phenyl-4h-chromen-4-one,crysin,4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl,unii-3cn01f5zj5,flavone, 5,7-dihydroxy,5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one,chembl117 PubChem CID: 5281607 ChEBI: CHEBI:75095 IUPAC Name: 5,7-dihydroxy-2-phenylchromen-4-one SMILES: C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O