Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysin, 99.4%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15498505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15498505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15498505 Supplier MP Biomedicals, Inc Supplier No. 0215498505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naturally occurring flavonoid used for research or further manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-40-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00006593
InChI Key RTIXKCRFFJGDFG-UHFFFAOYSA-N
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
PubChem CID 92930
IUPAC Name 5,7-dihydroxy-2-phenyl-4H-chromen-4-one
SMILES OC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1

Specifications

Color Yellow
Assay Percent Range 99.4%
Quantity 5 g
Formula Weight 254.24
Grade Research
Physical Form Powder
Chemical Name or Material Chrysin, For HPLC analysis
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.