CAS RN 480-41-1
IUPAC Name:
5,7-dihydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms:
asahina naringenine pelargidanon naringetol salipurpol salipurol narigenin
Molecular Weight (g/mol):
272.26
Molecular Formula:
C15H12O5
InChi Key:
FTVWIRXFELQLPI-UHFFFAOYNA-N
SMILES:
OC1=CC=C(C=C1)C1CC(=O)C2=C(O)C=C(O)C=C2O1
1
–
1
of
1
results