Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Naphthol As-Mx Phosphate Free Acid Naphthol-Free MP Biomedicals
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| CAS | 480-41-1 |
| Color | White |
| Molecular Formula | C15H12O5 |
| Quantity | 100 mg |
| Synonym | 2r-naringenin, r-naringenin, naringenin, 2r-5,7-dihydroxy-2-4-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, --naringenin, +-2r-5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, 2,3-dihydro-5,7-dihydroxy-2-4-hydroxyphenyl-4-benzopyrone, +-naringenin, 2r-5,7,4'-trihydroxyflavone, 2r-4',5,7-trihydroxyflavanone |
| InChI Key | FTVWIRXFELQLPI-CYBMUJFWSA-N |
| SMILES | C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O |
| IUPAC Name | (2R)-5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one |
| Molecular Weight (g/mol) | 272.256 |
| PubChem CID | 667495 |
| Show More |
Safety and Handling
| Recommended Storage | Store at -0°C. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?