accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOHOHO

CAS RN 480-67-1

IUPAC Name: 2,6-dihydroxy-4-methylbenzoic acid
Synonyms: 2,6-dihydroxy-4-methylbenzoic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: YBZAVRDNSPUMFK-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C(O)=O)C(O)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results