Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dihydroxy-4-methylbenzoic Acid, TCI America™
SDP

Catalog No. D27705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27705G Supplier TCI America Supplier No. D27705G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 480-67-1
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00184686
InChI Key YBZAVRDNSPUMFK-UHFFFAOYSA-N
Synonym 2,6-Dihydroxy-p-toluic Acid
PubChem CID 68074
IUPAC Name 2,6-dihydroxy-4-methylbenzoic acid
SMILES CC1=CC(=C(C(=C1)O)C(=O)O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 168.15
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dihydroxy-4-methylbenzoic Acid
RTECSNumber VH3709600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.