accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4801-80-3

IUPAC Name: benzyl N-(1-carbamoyl-2-phenylethyl)carbamate
Synonyms: benzyl N-(1-carbamoyl-2-phenylethyl)carbamate
Molecular Weight (g/mol): 298.34
Molecular Formula: C17H18N2O3
InChi Key: HHBOFAIEPRHUSR-UHFFFAOYNA-N
SMILES: NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results