accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOO

CAS RN 4802-47-5

IUPAC Name: 4,10-dioxatricyclo[6.3.0.0²,⁶]undecane-3,5,9,11-tetrone
Synonyms: 4,10-dioxatricyclo[6.3.0.0²,⁶]undecane-3,5,9,11-tetrone
Molecular Weight (g/mol): 210.14
Molecular Formula: C9H6O6
InChi Key: NLWBEORDOPDUPM-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2C1CC1C2C(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results