Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic dianhydride, Thermo Scientific™

Catalog No. AC111515000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111515000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111515000 Supplier Thermo Scientific Chemicals Supplier No. 111515000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4802-47-5
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00046872
InChI Key NLWBEORDOPDUPM-UHFFFAOYNA-N
Synonym cis-1,2,3,4-cyclopentanetetracarboxylic dianhydride, cyclopentane-1alpha,2alpha,3alpha,4alpha-tetracarboxylic acid 1,2:3,4-dianhydride
PubChem CID 2723616
IUPAC Name 4,10-dioxatricyclo[6.3.0.0²,⁶]undecane-3,5,9,11-tetrone
SMILES O=C1OC(=O)C2C1CC1C2C(=O)OC1=O

Specifications

Assay Percent Range 70%
Quantity 500 g
Formula Weight 210.14
Chemical Name or Material cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic dianhydride, tech., 70%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.