accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONMgBr

CAS RN 480424-75-7

IUPAC Name: 4-{[4-(bromomagnesio)phenyl]methyl}morpholine
Synonyms: 4-{[4-(bromomagnesio)phenyl]methyl}morpholine
Molecular Weight (g/mol): 280.45
Molecular Formula: C11H14BrMgNO
InChi Key: OFPHGAKUXVPRKI-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=C(CN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results