accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 480425-35-2

IUPAC Name: methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 262.11
Molecular Formula: C14H19BO4
InChi Key: JBJGSVBGUBATNH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results