Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98.0+%, TCI America™
SDP

Catalog No. M26655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26655G 5 g
M26651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26655G Supplier TCI America Supplier No. M26655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 480425-35-2
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.112
MDL Number MFCD03789258
InChI Key JBJGSVBGUBATNH-UHFFFAOYSA-N
Synonym 3-methoxycarbonylphenylboronic acid pinacol ester, methyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-methoxycarbonylphenylboronic acid, pinacol ester, 3-methoxycarbonyl phenylboronic acid pinacol ester, methyl 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, pubchem9527, 3-carbomethoxyphenylboronic acid pinacol ester, 3-methoxycarbonyl phenyl boronic acid pinacol ester
PubChem CID 2734651
IUPAC Name methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)OC

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 262.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.