accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)OHO(S)HNH3COOOCH3CH3CH3H3C

CAS RN 48068-25-3

IUPAC Name: (2S,3R)-2-{[(tert-butoxy)carbonyl]amino}-3-methoxybutanoic acid
Synonyms: (2S,3R)-2-[(tert-butoxycarbonyl)amino]-3-methoxybutanoic acid|BOC-O-METHYL-L-THREONINE
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: VWSUOKFUIPMDDX-RQJHMYQMSA-N
SMILES: CO[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results