Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-O-methyl-L-threonine, 95%, Thermo Scientific™

Catalog No. AAH6375006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6375006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6375006 Supplier Thermo Scientific Chemicals Supplier No. H6375006

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48068-25-3
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00076990
InChI Key VWSUOKFUIPMDDX-RQJHMYQMSA-N
Synonym boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
PubChem CID 15460346
IUPAC Name (2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CO[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 5 g
Formula Weight 233.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-O-methyl-L-threonine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.