CAS RN 48150-45-4
IUPAC Name:
[3-(2-methylprop-2-enamido)phenyl]boronic acid
Synonyms:
3-(2-methylprop-2-enamido)phenylboronic acid
Molecular Weight (g/mol):
205.02
Molecular Formula:
C10H12BNO3
InChi Key:
GBBUBIKYAQLESK-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)NC1=CC=CC(=C1)B(O)O
1
–
2
of
2
results
2
Sigma Aldrich 3-(Methacryloylamino)phenylboronic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.