Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methacrylamidophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M26765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2676-5G 5 g
M26761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2676-5G Supplier TCI America Supplier No. M26765G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 48150-45-4
Color Yellow
Molecular Formula C10H12BNO3
Quantity 5 g
MDL Number MFCD09475805
Synonym 3-Methacrylamidobenzeneboronic Acid
InChI Key GBBUBIKYAQLESK-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)NC(=O)C(=C)C)(O)O
IUPAC Name [3-(2-methylprop-2-enoylamino)phenyl]boronic acid
Molecular Weight (g/mol) 205.02
PubChem CID 3016474
Formula Weight 205.02
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.