accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3OO

CAS RN 482-27-9

IUPAC Name: 4,9-dimethoxy-7H-furo[3,2-g]chromen-7-one
Synonyms: isopimpinellin 5,8-dimethoxypsoralen 5,8-dimethoxypsoralene
Molecular Weight (g/mol): 246.22
Molecular Formula: C13H10O5
InChi Key: DFMAXQKDIGCMTL-UHFFFAOYSA-N
SMILES: COC1=C2OC=CC2=C(OC)C2=C1OC(=O)C=C2
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results