accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 482-44-0

IUPAC Name: 9-[(3-methylbut-2-en-1-yl)oxy]-7H-furo[3,2-g]chromen-7-one
Synonyms: imperatorin marmelosin pentosalen ammidin 8-isoamylenoxypsoralen 8-isopentenyloxypsoralene
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: OLOOJGVNMBJLLR-UHFFFAOYSA-N
SMILES: CC(C)=CCOC1=C2OC=CC2=CC2=C1OC(=O)C=C2
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)