accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 482-74-6

CH3ONCH3OOCH3OO

CAS RN 482-74-6

IUPAC Name: 6,7-dimethoxy-12-methyl-16,18-dioxa-12-azatetracyclo[12.7.0.0⁴,⁹.0¹⁵,¹⁹]henicosa-1(14),4(9),5,7,15(19),20-hexaen-3-one
Synonyms: cryptopine
Molecular Weight (g/mol): 369.42
Molecular Formula: C21H23NO5
InChi Key: XPOJSWHIKCNLEQ-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1OC)C(=O)CC1=C(CN(C)CC2)C2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results