Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryptopine, MP Biomedicals

Catalog No. ICN201142 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201142 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201142 Supplier MP Biomedicals, Inc Supplier No. 0520114201

Chemical Identifiers

CAS 482-74-6
Molecular Formula C21H23NO5
Molecular Weight (g/mol) 369.417
InChI Key XPOJSWHIKCNLEQ-UHFFFAOYSA-N
Synonym cryptopine, cryptocavine, cryptopin, kryptocavin, kryptopine, unii-mw13x5yk4a, mw13x5yk4a, 12,13,14,15,-tetrahydro-9,10-dimethoxy-14-methylbenzo e-1,3-dioxolo 4,5-1 2 benzazecin-7 6h-one, benzo e-1,3-dioxolo 4,5-l 2 benzazecin-12 5h-one, 4,6,7,13-tetrahydro-9,10-dimethoxy-5-methyl, 4-27-00-06652 beilstein handbook reference
PubChem CID 72616
SMILES CN1CCC2=CC(=C(C=C2C(=O)CC3=C(C1)C4=C(C=C3)OCO4)OC)OC

Specifications

Quantity 1 g
Formula Weight 369.4
Chemical Name or Material Cryptopine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.