accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 483-84-1

IUPAC Name: 8-hydroxy-5,7-dinitronaphthalene-2-sulfonic acid
Synonyms: naphthol yellow S
Molecular Weight (g/mol): 314.22
Molecular Formula: C10H6N2O8S
InChi Key: FCQJEPASRCXVCB-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C2=CC=C(C=C12)S(O)(=O)=O)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results