Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavianic Acid Dihydrate 98.0+%, TCI America™
SDP

Catalog No. F001225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F001225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F001225G Supplier TCI America Supplier No. F001225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 483-84-1
Molecular Formula C10H6N2O8S
Molecular Weight (g/mol) 314.224
MDL Number MFCD00046407
InChI Key FCQJEPASRCXVCB-UHFFFAOYSA-N
Synonym 2,4-Dinitro-1-naphthol-7-sulfonic Acid, 8-Hydroxy-5,7-dinitro-2-naphthalenesulfonic Acid
PubChem CID 10226
ChEBI CHEBI:87221
IUPAC Name 8-hydroxy-5,7-dinitronaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=C1S(=O)(=O)O)C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])O

Specifications

Melting Point 149°C
Color Yellow
Quantity 25 g
UN Number 3261
Formula Weight 314.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Flavianic Acid Dihydrate
RTECSNumber QK1810000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.