accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 484-20-8

IUPAC Name: 4-methoxy-7H-furo[3,2-g]chromen-7-one
Synonyms: bergapten 5-methoxyfuranocoumarin O-methylbergaptol 5-methoxy-6,7-furanocoumarin pentaderm bergapten(e) 5-methoxypsoralene 5-mop heraclin majudin
Molecular Weight (g/mol): 216.19
Molecular Formula: C12H8O4
InChi Key: BGEBZHIAGXMEMV-UHFFFAOYSA-N
SMILES: COC1=C2C=COC2=CC2=C1C=CC(=O)O2
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)