accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 484-65-1

IUPAC Name: 1-(chloromethyl)-2,3,4,5,6-pentamethylbenzene
Synonyms: 1-(chloromethyl)-2,3,4,5,6-pentamethylbenzene
Molecular Weight (g/mol): 196.72
Molecular Formula: C12H17Cl
InChi Key: CXUAEBDTJFKMBV-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(CCl)C(C)=C1C
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results