Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentamethylbenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. P20465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20465G 5 g
P204625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20465G Supplier TCI America Supplier No. P20465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 484-65-1
Molecular Formula C12H17Cl
Molecular Weight (g/mol) 196.718
MDL Number MFCD00000900
InChI Key CXUAEBDTJFKMBV-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentamethylbenzyl chloride, pentamethylbenzyl chloride, benzene, chloromethyl pentamethyl, chloromethyl pentamethylbenzene, 1-chloromethyl-2,3,4,5,6-pentamethylbenzene, benzene, 1-chloromethyl-2,3,4,5,6-pentamethyl, acmc-209kd7, cxuaebdtjfkmbv-uhfffaoysa, 2,3,4,6-pentamethylbenzyl chloride, 2,4,5,6-pentamethylbenzyl chloride
PubChem CID 68087
IUPAC Name 1-(chloromethyl)-2,3,4,5,6-pentamethylbenzene
SMILES CC1=C(C(=C(C(=C1C)C)CCl)C)C

Specifications

Melting Point 82°C
Color White-Yellow
Boiling Point 163°C
Quantity 5 g
UN Number 3261
Formula Weight 196.72
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.