accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4841-20-7

IUPAC Name: methyl 2-(2,4,5-trichlorophenoxy)propanoate
Synonyms: methyl 2-(2,4,5-trichlorophenoxy)propanoate
Molecular Weight (g/mol): 283.53
Molecular Formula: C10H9Cl3O3
InChi Key: YTAXYXOJOYIQQO-UHFFFAOYNA-N
SMILES: COC(=O)C(C)OC1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results