accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHNN

CAS RN 4856-97-7

IUPAC Name: (1H-1,3-benzodiazol-2-yl)methanol
Synonyms: 1H-1,3-benzodiazol-2-ylmethanol
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: IAJLTMBBAVVMQO-UHFFFAOYSA-N
SMILES: OCC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results