Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Hydroxymethyl)benzimidazole 98.0+%, TCI America™
SDP

Catalog No. H12295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12295G Supplier TCI America Supplier No. H12295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4856-97-7
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD00014560
InChI Key IAJLTMBBAVVMQO-UHFFFAOYSA-N
Synonym 1h-benzimidazole-2-methanol, 1h-benzoimidazol-2-yl methanol, 2-benzimidazolemethanol, 2-hydroxymethyl benzimidazole, 1h-benzo d imidazol-2-yl methanol, 2-hydroxymethylbenzimidazole, 2-hydroxymethyl-1h-benzimidazole, 1h-benzoimidazol-2-yl-methanol, 1h-1,3-benzodiazol-2-ylmethanol, 1h-benzimidazol-2-yl methanol
PubChem CID 78569
IUPAC Name 1H-benzimidazol-2-ylmethanol
SMILES C1=CC=C2C(=C1)NC(=N2)CO

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 5 g
Formula Weight 148.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Hydroxymethyl)benzimidazole
EINECSNumber (5)-0469
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.