CAS RN 486-66-8
4',7-Dihydroxyisoflavone, 98+%
CAS: 486-66-8 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.241 MDL Number: MFCD00016954 InChI Key: ZQSIJRDFPHDXIC-UHFFFAOYSA-N Synonym: daidzein,4',7-dihydroxyisoflavone,daidzeol,7,4'-dihydroxyisoflavone,7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one,diadzein,7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl,7-hydroxy-3-4-hydroxyphenyl chromen-4-one,7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone PubChem CID: 5281708 ChEBI: CHEBI:28197 IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one SMILES: C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
Daidzein, 98%
CAS: 486-66-8 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.24 MDL Number: MFCD00016954 InChI Key: ZQSIJRDFPHDXIC-UHFFFAOYSA-N Synonym: daidzein,4',7-dihydroxyisoflavone,daidzeol,7,4'-dihydroxyisoflavone,7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one,diadzein,7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl,7-hydroxy-3-4-hydroxyphenyl chromen-4-one,7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone PubChem CID: 5281708 ChEBI: CHEBI:28197 IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one SMILES: C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
4',7-Dihydroxyisoflavone, 97%
CAS: 486-66-8 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.241 MDL Number: MFCD00016954 InChI Key: ZQSIJRDFPHDXIC-UHFFFAOYSA-N Synonym: daidzein,4',7-dihydroxyisoflavone,daidzeol,7,4'-dihydroxyisoflavone,7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one,diadzein,7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl,7-hydroxy-3-4-hydroxyphenyl chromen-4-one,7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone PubChem CID: 5281708 ChEBI: CHEBI:28197 IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one SMILES: C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
Daidzein Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
MilliporeSigma™ Calbiochem™ Daidzein
Inactive analog of Genistein.
Daidzein, 98.7%, MP Biomedicals
CAS: 486-66-8 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.241 InChI Key: ZQSIJRDFPHDXIC-UHFFFAOYSA-N Synonym: daidzein,4',7-dihydroxyisoflavone,daidzeol,7,4'-dihydroxyisoflavone,7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one,diadzein,7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl,7-hydroxy-3-4-hydroxyphenyl chromen-4-one,7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone PubChem CID: 5281708 ChEBI: CHEBI:28197 IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one SMILES: C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
Daidzein, ≥97% (HPLC), MilliporeSigma™ Supelco™
Daidzein is an endocrine-active estrogenic isoflavone. This phytoestrogen is capable to bind to estrogen to exert estrogenic effects in vivo. It can naturally be found in soybean seed and flour.
Daidzein 98.0+%, TCI America™
CAS: 486-66-8 Molecular Formula: C15H10O4 Molecular Weight (g/mol): 254.241 MDL Number: MFCD00016954 InChI Key: ZQSIJRDFPHDXIC-UHFFFAOYSA-N Synonym: daidzein,4',7-dihydroxyisoflavone,daidzeol,7,4'-dihydroxyisoflavone,7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one,diadzein,7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl,7-hydroxy-3-4-hydroxyphenyl chromen-4-one,7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone PubChem CID: 5281708 ChEBI: CHEBI:28197 IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one SMILES: C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O