accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 486424-37-7

IUPAC Name: 3-amino-6-bromopyrazine-2-carboxylic acid
Synonyms: 3-amino-6-bromopyrazine-2-carboxylic acid
Molecular Weight (g/mol): 218.01
Molecular Formula: C5H4BrN3O2
InChi Key: MTNAQEKMSVDTAQ-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)N=C1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results