Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-6-bromopyrazine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH6690806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6690806 5 g
AAH6690803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6690806 Supplier Thermo Scientific Chemicals Supplier No. H6690806

missing translation for 'validMainframeMsg'

3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486424-37-7
Molecular Formula C5H4BrN3O2
Molecular Weight (g/mol) 218.01
MDL Number MFCD08275681
InChI Key MTNAQEKMSVDTAQ-UHFFFAOYSA-N
Synonym 3-amino-6-bromo-pyrazine-2-carboxylic acid, 3-amino-6-bromo-2-pyrazinecarboxylic acid, 2-amino-5-bromopyrazine-3-carboxylic acid, pyrazinecarboxylic acid, 3-amino-6-bromo, pubchem22388, 3-amino-6-bromopyrazine-2-carboxylicacid, acmc-1bna1, ksc235o0r, 2-pyrazinecarboxylic acid, 3-amino-6-bromo, 3-azanyl-6-bromanyl-pyrazine-2-carboxylic acid
PubChem CID 18521600
IUPAC Name 3-amino-6-bromopyrazine-2-carboxylic acid
SMILES C1=C(N=C(C(=N1)N)C(=O)O)Br

Specifications

Melting Point 178°C to 183°C
Color Yellow
Quantity 5 g
Beilstein 147851
Sensitivity Light sensitive
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 218.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-6-bromopyrazine-2-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.