CAS RN 4869-46-9
CAS RN 4869-46-9
IUPAC Name:
1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde
Synonyms:
1,3-dimethyl-2,4-dioxopyrimidine-5-carbaldehyde
Molecular Weight (g/mol):
168.15
Molecular Formula:
C7H8N2O3
InChi Key:
KGJFRZOATIXYPW-UHFFFAOYSA-N
SMILES:
CN1C=C(C=O)C(=O)N(C)C1=O
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 1,3-dimethyluracil-5-carboxaldehyde
Sigma Aldrich 1,3-dimethyluracil-5-carboxaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.