accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 487-03-6

IUPAC Name: 1-methyl-2H,7H,9H-pyrido[3,4-b]indol-7-one
Synonyms: harmol
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: LBBJNGFCXDOYMQ-UHFFFAOYSA-N
SMILES: CC1=C2NC3=CC(=O)C=CC3=C2C=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results