Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmol 98.0+%, TCI America™
SDP

Catalog No. H1360200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1360200MG 200 mg
H13601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1360200MG Supplier TCI America Supplier No. H1360200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 487-03-6
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.225
MDL Number MFCD00834164
InChI Key LBBJNGFCXDOYMQ-UHFFFAOYSA-N
Synonym 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol
PubChem CID 68094
IUPAC Name 1-methyl-2,9-dihydropyrido[3,4-b]indol-7-one
SMILES CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1

Specifications

Melting Point 323°C
Color Yellow
Quantity 200 mg
Formula Weight 198.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Harmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.