accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHNOSN

CAS RN 487020-03-1

IUPAC Name: N-{12-cyanoindolizino[2,3-b]quinoxalin-2-yl}thiophene-2-carboxamide
Synonyms: N-{12-cyanoindolizino[2,3-b]quinoxalin-2-yl}thiophene-2-carboxamide
Molecular Weight (g/mol): 369.4
Molecular Formula: C20H11N5OS
InChi Key: BCSBXWKRZUPFHW-UHFFFAOYSA-N
SMILES: O=C(NC1=CC2=C(C#N)C3=NC4=CC=CC=C4N=C3N2C=C1)C1=CC=CS1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results